(±)-α-Lipoic acid, high purity

Supplier: VWR Chemicals
Warning

Synonyms: 5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)​-​1,2-​Dithiolane-​3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid

VWRC0494-25GEA 221 SGD
VWRC0494-25G
(±)-α-Lipoic acid, high purity
(±)-α-Lipoic acid
Formula: C₈H₁₄O₂S₂
MW: 206.33 g/mol
Boiling Pt: 160 – 165 °C
Melting Pt: 59…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00005474
CAS Number: 1077-28-7

Order Now

Specification Test Results

Identification (IR) PASS
Purity ≥ 99.0 %
Solubility (5%, EtOH) PASS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR