Polyvinylpyrrolidone, M.W. ∼40000 99, high_purity (K30)

Supplier: VWR Chemicals

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

VWRC0507-500GEA 268 SGD
VWRC0507-500G
Polyvinylpyrrolidone, M.W. ∼40000 99, high_purity (K30)
Polyvinylpyrrolidone

Common reagent for nucleic acid hybridisation procedures for detection of DNA/RNA.

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7893e4d0affb7fb76c948d932b4a0e880dece26c&vid=7893e4d0affb7fb76c948d932b4a0e880dece26c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Heavy Metals <0.001%
pH (5%, Water) @25 °C 3 – 7
Purity (Dry Basis) >99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR