Sodium acetate trihydrate 99-101 ACS

Supplier: VWR Chemicals

Synonyms: Acetic acid sodium salt trihydrate

VWRC0530-1KGEA 176 SGD
VWRC0530-1KG VWRC0530-2.5KG VWRC0530-500G
Sodium acetate trihydrate 99-101 ACS
Sodium acetate trihydrate
Formula: H₃CCOONa·3H₂O
MW: 136.08 g/mol
Boiling Pt: 120 – 123 °C (1013 hPa)
Melting Pt: 58
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071557
CAS Number: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Calcium <0.005%
Chloride ≤0.001%
Heavy Metals (as Pb) ≤0.0005%
Insolubles ≤0.005%
Iron ≤0.0005%
Magnesium ≤0.002%
Permanganate Reducers Test Pass
pH (5%, Water) @25 °C 7.5 - 9.2
Phosphate ≤0.0005%
Potassium ≤0.005%
Purity 99 – 101%
Sulphate ≤0.002%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR