Bromocresol green sodium salt ACS

Supplier: VWR Chemicals

Synonyms: Bromocresol green water soluble, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-2,2-dioxido-3H-benzo[d][1,2]oxathiol-3-yl)-3-methylphenolate

VWRC0548-50GEA 462 SGD
VWRC0548-50G
Bromocresol green sodium salt ACS
Bromocresol green sodium salt

A pH indicator (yellow to blue; range 3,8 to 5,4).

Formula: C₂₁H₁₃Br₄NaO₅S
MW: 720 g/mol
Melting Pt: 225
Storage Temperature: Ambient
MDL Number: MFCD00148898
CAS Number: 62625-32-5
EINECS: 263-657-4

Order Now

Specification Test Results

Insolubles Test none detected
Visual Transition Interval (pH 3.8 – 5.4) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR