Sodium carbonate, anhydrous 99,5 ACS

Supplier: VWR Chemicals
Warning

Synonyms: Calcined soda, Carbonic acid disodium salt

VWRC0585-500GEA 192 SGD
VWRC0585-500G
Sodium carbonate, anhydrous 99,5 ACS
Sodium carbonate
Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600
Melting Pt: 854
Density: 2.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Calcium <0.03%
Chloride ≤0.001%
Heavy metals (as Pb) ≤0.0005%
Insolubles ≤0.01%
Iron ≤0.0005%
Loss on heating ≤1%
Magnesium ≤0.005%
Phosphate ≤0.001%
Potassium ≤0.005%
Purity ≥99.5%
Silica ≤0.005%
Sulphur compounds ≤0.003%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR