Iron(III) sulphate hydrate, Reagent Grade

Supplier: VWR Chemicals
Warning

Synonyms: Ferric Persulphate, Diiron trissulphate hydrate, Ferric sulphate hydrate

VWRC0629-500GEA 297 SGD
VWRC0629-500G
Iron(III) sulphate hydrate, Reagent Grade
Iron (III) sulphate hydrate
Formula: Fe₂(SO₄)₃·xH₂O
MW: 399.88 g/mol (anhy)
Density: 3.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149714
CAS Number: 15244-10-7
EINECS: 233-072-9
Merck Index: 13,04065

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR