Phenol red sodium salt ACS

Supplier: VWR Chemicals

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide

VWRC0648-50GEA 250 SGD
VWRC0648-50G
Phenol red sodium salt ACS
Phenol red sodium salt
Formula: C₁₉H₁₃NaO₅S
Melting Pt: 285 °C
Storage Temperature: Ambient
MDL Number: MFCD00066901
CAS Number: 34487-61-1
EINECS: 252-057-8
Merck Index: 12,07397

Order Now

Specification Test Results

Solubility (0.1%, water) Pass
Visual transition interval (pH 6.8 – 8.2) Pass

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR