DL-Malic acid 99-100.5%, high purity

Supplier: VWR Chemicals
Danger

Synonyms: DL-Hydroxysuccinic acid, (±)-Malic acid, (±)-Hydroxy-1,4-butanedioic acid, (±)-Hydroxysuccinic acid, Hydroxybutanedioic acid, 2-Hydroxysuccinic acid

VWRC0664-2.5KGEA 676 SGD
VWRC0664-2.5KG
DL-Malic acid 99-100.5%, high purity
DL-Malic acid
Formula: HOOCCH(OH)CH₂COOH
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 131…132 °C
Density: 1,601 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064212
CAS Number: 617-48-1
EINECS: 210-514-9
Merck Index: 13,05730
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=920b54632a869bc5df22624fb9fecd4d11cae65e&vid=278193678126274041defdc0bd71c7c40efb3085&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Fumaric Acid <1%
Heavy Metals (as Pb) ≤0.001%
Insolubles ≤0.1%
Maleic Acid ≤0.05%
Purity 99 – 100.5%
Residue on Ignition ≤0.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR