Iron(III) chloride hexahydrate 97-102% ACS

Supplier: VWR Chemicals
Danger

Synonyms: Iron trichloride hexahydrate, Ferric chloride hexahydrate

VWRC0682-1KGEA 289 SGD
VWRC0682-1KG VWRC0682-2.5KG VWRC0682-500G
Iron(III) chloride hexahydrate 97-102% ACS
Iron (III) chloride hexahydrate
Formula: FeCl₃·6H₂O
MW: 270.3 g/mol
Boiling Pt: 280 – 285 °C (1013 hPa)
Melting Pt: 37
Density: 1.82 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00149712
CAS Number: 10025-77-1
EINECS: 231-729-4
UN: 3260
ADR: 8 III
Merck Index: 13,04048
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=808c75376e55f29296d58eb1e5a1e184039ce733&vid=6d820873011bee3ff401441544f7f85599e40626&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Calcium <0.01%
Copper ≤0.003%
Ferrous Iron ≤0.002%
Insolubles ≤0.01%
Magnesium ≤ 0.005%
Nitrate ≤0.01%
Phosphorus Compounds (as PO₄) ≤0.01%
Potassium ≤0.005%
Purity 97 – 102%
Sodium ≤0.05%
Sulphate ≤0.01%
Zinc ≤0.003%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR