EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥97%, white crystalline powder, Ultra Pure Grade

Supplier: VWR Chemicals

Synonyms: EGTA, Ethylenebis(oxyethylenenitrilo)tetra(acetic acid), Kestranal® FE PA, Idranal® VI, Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, Ethylenebis(oxyethylenenitrilo)tetraacetic acid, Ethylene Glycol Bis(2-aminoethyl Ether)-N,N,N,N-tetraacetic acid, 3,12-Bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecanedioic acid, Titriplex® VI

VWRC0732-10GEA 187 SGD
VWRC0732-10G VWRC0732-100G
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥97%, white crystalline powder, Ultra Pure Grade
EGTA (Ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid)
Formula: C₁₄H₂₄N₂O₁₀
Melting Pt: 241 °C
Storage Temperature: Ambient
MDL Number: MFCD00004291
CAS Number: 67-42-5
EINECS: 200-651-2
Merck Index: 13,03560

Order Now

Specification Test Results

DNase none detected
Heavy Metals (as Pb) ≤0.001%
Loss on Drying ≤1%
Melting Range w/Decomposition ≥238 °C
Protease none
Purity >97 %
RNase none detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR