Lithium dodecyl sulphate ≥99%, high purity

Supplier: VWR Chemicals
Warning

VWRC0782-100GEA 486 SGD
VWRC0782-100G VWRC0782-25G
Lithium dodecyl sulphate ≥99%, high purity
Lithium dodecyl sulphate
Formula: C₁₂H₂₅LiO₄S
Storage Temperature: Ambient
MDL Number: MFCD00007467
CAS Number: 2044-56-6
EINECS: 218-058-2
UN: 1325
ADR: 4.1,III

Order Now

Specification Test Results

Abs. at 260nm (5%, Water) ≤0.1
Abs.at 280nm (5%, Water) ≤0.1
Loss on drying ≤3%
Purity ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR