Sodium sulphate, anhydrous 99 ACS

Supplier: VWR Chemicals

Synonyms: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt

VWRC0836-1KGEA 151 SGD
VWRC0836-1KG VWRC0836-2.5KG VWRC0836-500G
Sodium sulphate, anhydrous 99 ACS
Sodium sulphate
Formula: Na₂SO₄
MW: 142.04 g/mol
Boiling Pt: 1429
Melting Pt: 888
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Calcium <0.01%
Chloride ≤0.001%
Heavy metals (as Pb) ≤0.0005%
Insolubles ≤0.01%
Iron ≤0.001%
Loss on ignition ≤0.5%
Magnesium ≤0.005%
Nitrogen compounds ≤0.0005%
pH (5%, Water) at 25 °C 5.2 – 9.2
Phosphate ≤0.001%
Potassium ≤0.01%
Purity ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR