Sodium dodecyl sulphate (SDS) 20% aqueous solution Biotechnology

Supplier: VWR Chemicals
Warning

Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

VWRC0837-200MLEA 124 SGD
VWRC0837-200ML VWRC0837-500ML
Sodium dodecyl sulphate (SDS) 20% aqueous solution Biotechnology
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Density: 1…1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Conductivity 22800 umhos - 27600 umhos
pH @25 °C 5 - 8


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR