Formic acid ≥94.5%, high purity

Supplier: VWR Chemicals
Danger

VWRC0961-100MLEA 183 SGD
VWRC0961-100ML VWRC0961-500ML
Formic acid ≥94.5%, high purity
Formic acid
Formula: HCO₂H
Boiling Pt: 100…101 °C (1013 hPa)
Melting Pt: 8,2…8.4 °C
Density: 1,22 g/cm³ (25 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8 (3),II
REACH: 01-2119491174-37
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥94.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR