tetra-Chloroauric(III) acid trihydrate, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Danger

VWRC1028.0001EA 606 SGD
VWRC1028.0001
tetra-Chloroauric(III) acid trihydrate, AnalaR NORMAPUR® analytical reagent
tetra-Chloroauric (III) acid trihydrate
Formula: HAuCl₄·3H₂O
Melting Pt: 30 °C
Density: 3,9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149904
CAS Number: 16961-25-4
EINECS: 240-948-4
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay (Au) Min. 48 %
Alkali metals Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR