Chlorobenzene ≥99.5%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Warning

Synonyms: Phenylchloride, Benzene Chloride, Monochlorobenzene

VWRC103386DEA 274 SGD
VWRC103386D
Chlorobenzene ≥99.5%, AnalaR NORMAPUR® analytical reagent
Chlorobenzene
Formula: C₆H₅Cl
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1,11 g/cm³ (20 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3,III
REACH: 01-2119432722-45
Merck Index: 13,02139
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.5 %
IR Spectrum Passes test
Colouration Max. 10 APHA
Density (20/4) 1.100 - 1.110
Evaporation residue Max. 0.05 %
Water Max. 0.05 %
Al (Aluminium) Max. 2.0 ppm
Ba (Barium) Max. 1.0 ppm
Ca (Calcium) Max. 5.0 ppm
Cd (Cadmium) Max. 0.5 ppm
Co (Cobalt) Max. 0.2 ppm
Cr (Chromium) Max. 0.2 ppm
Cu (Copper) Max. 0.2 ppm
Fe (Iron) Max. 2.0 ppm
K (Potassium) Max. 1.0 ppm
Mg (Magnesium) Max. 0.5 ppm
Mn (Manganese) Max. 0.2 ppm
Na (Sodium) Max. 5.0 ppm
Ni (Nickel) Max. 0.2 ppm
Pb (Lead) Max. 0.5 ppm
Sr (Strontium) Max. 0.2 ppm
Zn (Zinc) Max. 1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR