Lanthanum(III) chloride heptahydrate 98, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals

VWRC103433QEA 85.7 SGD
VWRC103433Q
Lanthanum(III) chloride heptahydrate 98, AnalaR NORMAPUR® zA
Lanthanum (III) chloride heptahydrate
Formula: LaCl₃·7H₂O
MW: 371.37 g/mol
Boiling Pt: < 860 °C (1013 hPa)
Melting Pt: 96
Storage Temperature: Ambient
MDL Number: MFCD00149756
CAS Number: 10025-84-0
EINECS: 233-237-5

Order Now

Specification Test Results

Assay Min. 98 %
Identification Passes test
Insolubility in water Max. 0.1 %
Ca (Calcium) Max. 10 ppm
Cu (Copper) Max. 1 ppm
Fe (Iron) Max. 5 ppm
Mg (Magnesium) Max. 1 ppm
Pb (Lead) Max. 2 ppm
Zn (Zinc) Max. 3 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR