Sodium benzoate ≥99.5%, AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Warning

Synonyms: Benzoic acid sodium salt

VWRC103974REA 65.4 SGD
VWRC103974R
Sodium benzoate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Sodium benzoate
Formula: C₆H₅COONa
MW: 144.11 g/mol
Melting Pt: 410…430 °C
Density: 1.44 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS Number: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 13,08654

Order Now

Specification Test Results

Assay (calculated on dried substance) Min. 99.5 %
pH (20°C; 5 %) 7 - 9
Insolubility in water Max. 50 ppm
Loss on drying (100°C) Max. 1.0 %
Total Cl (Chlorine) Max. 0.03 %
Cl (Chloride) Max. 50 ppm
SO4 (Sulphate) Max. 100 ppm
Ca (Calcium) Max. 100 ppm
Cu (Copper) Max. 1 ppm
Fe (Iron) Max. 1 ppm
K (Potassium) Max. 0.05 %
Mg (Magnesium) Max. 50 ppm
Pb (Lead) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR