Sodium dodecyl sulphate (SDS) 99, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals
Danger

Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

VWRC108073JEA 76.3 SGD
VWRC108073J
Sodium dodecyl sulphate (SDS) 99, AnalaR NORMAPUR® zA
Sodium dodecyl sulphate (SDS)

A general biochemical reagent often used as a detergent for electrophoresis

Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Melting Pt: 204…207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 100 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1 III
REACH: 01-2119489461-32
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated on anhydrous) Min. 99 %
Assay (C12 compounds)(calc.on anhydrous) Min. 99 %
IR Spectrum Passes test
Insolubility in water Max. 30 ppm
Loss on drying (100°C) Max. 1.5 %
Cl (Chloride) Max. 100 ppm
PO4 (Phosphate) Max. 10 ppm
Cu (Copper) Max. 1 ppm
Fe (Iron) Max. 2 ppm
Pb (Lead) Max. 5 ppm
Absorbance (220-350 nm) (3 %) Max. 0.1

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR