VWR®, Chlorpyrifos, pesticide reference standard, 100 µg/ml in methanol

Supplier: VWR Chemicals

VWRC123692UEA 97.6 SGD
VWRC123692U
VWR®, Chlorpyrifos, pesticide reference standard, 100 µg/ml in methanol
Standards Organic Standards

Order Now

SPECIFICATIONS

Description Chlorpyrifos
Environmentally Preferable
CAS No. 2921-88-2
Formula C₉H₁₁Cl₃NO₃PS

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR