VWR®, α-Endosulfan Certified Reference Material

Supplier: VWR Chemicals

VWRC124082EEA 144 SGD
VWRC124082E
VWR®, α-Endosulfan Certified Reference Material
Standards Organic Standards

Order Now

SPECIFICATIONS

Description α-Endosulfan
Environmentally Preferable
CAS No. 959-98-8
Formula C₉H₆Cl₆O₃S
Molecular weight 406.93 g/mol
Storage conditions Ambient (>5 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR