VWR®, Propyzamide neat Certified Reference Material

Supplier: VWR Chemicals

VWRC124482QEA 97.6 SGD
VWRC124482Q
VWR®, Propyzamide neat Certified Reference Material
Standards Organic Standards

Order Now

SPECIFICATIONS

Description Propyzamide
Environmentally Preferable
CAS No. 23950-58-5
MDL No. MFCD00055346
Formula C₁₂H₁₁Cl₂NO
Molecular weight 256.13 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR