Simazine(2-chloro-4,6-bis(ethylamino)-1,3,5-triazine) 99,5

Supplier: VWR Chemicals
Warning

VWRC124502DEA 97.6 SGD
VWRC124502D
Simazine(2-chloro-4,6-bis(ethylamino)-1,3,5-triazine) 99,5
Simazine (2-Chloro-4,6-bis(ethylamino)-1,3,5-triazine)
Formula: C₇H₁₂ClN₅
MW: 201.66 g/mol
Flash Pt: 100 °C (closed cup)
MDL Number: MFCD00023174
CAS Number: 122-34-9
EINECS: 204-535-2
UN: 3077

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR