VWR®, Organic reference standard, Phenol Mix 1, EPA 604
Supplier: VWR Chemicals
VWRC124742REA 163 SGD
VWRC124742R
VWR®, Organic reference standard, Phenol Mix 1, EPA 604
Standards Organic Standards
0,5 mg/ml of each component in methanol
2,4-Dichlorophenol ; 2,4-Dimethylphenol ; 2,4-Dinitrophenol ; 4-Chloro-3-methylphenol; 2-Chlorophenol ; 2-Methyl-4,6-dinitrophenol ; 2-Nitrophenol ; 4-Nitrophenol ; Phenol; Pentachlorophenol; 2,4,6-Trichlorophenol