Pesticides Mix 2, EPA 608
Supplier: VWR Chemicals
VWRC124782CEA 97.6 SGD
VWRC124782C
Pesticides Mix 2, EPA 608
Standards Organic Standards
Chlordane (0,02 mg/ml) and Toxaphene (0,20 mg/ml) in methanol.