Peqlab peqGOLD, RNA Isolation, Total RNA Kit
Supplier: VWR Chemicals
VWRC13-6834-02EA 677 SGD
VWRC13-6834-02
Peqlab peqGOLD, RNA Isolation, Total RNA Kit
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Easy to use method for isolation up to 100 µg of total RNA from cultured eukaryotic cells and soft tissues <1×10⁷ cells or 30 mg tissue.
  • Fast, 20 minutes or less
  • Safe with no phenol/chloroform extractions
  • Versatile with both spin and vacuum protocols available
  • Homogeniser column reduces RNA column clogging
  • Compatible with peqGOLD DNase I Digestion Kit for lowest level of gDNA co-purification (13-1091-01)
Delivery information: See attached manual.