Bottles, wide neck, square for solid products
Supplier: VWR Chemicals
VWRC13251000EA 2 SGD
VWRC13251000
Bottles, wide neck, square for solid products
Bottles Plastic Bottles
PE.
  • Ideal for storing and shipping samples
  • Wide mouth permits easy filling, emptying and cleaning
  • Square shape saves space
Order Now
SPECIFICATIONS
Environmentally Preferable