n-Hexane ≥99.0%, SPECTRONORM® for spectroscopy

Supplier: VWR Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

Filtered through a 0.2 µm filter, packaged under nitrogen.

VWRC140096EEA 296 SGD
VWRC140096E
n-Hexane ≥99.0%, SPECTRONORM® for spectroscopy
n-Hexane

SPECTRONORM solvents have extensive specifications to meet the highly demanding requirements of UV applications.


  • Meets the requirements of "Specific use spectroscopy" according Reag. Ph.Eur.

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: –94,3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
REACH: 01-2119480412-44
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) ≥ 99.0 %
Acidity ≤ 0.0002 meq/g
Alkalinity ≤ 0.0002 meq/g
Colouration ≤ 10 APHA
Evaporation residue ≤ 2 ppm
Water ≤ 50 ppm
Absorbance (195 nm) ≤ 1.00
Absorbance (210 nm) ≤ 0.22
Absorbance (217 nm) ≤ 0.10
Absorbance (225 nm) ≤ 0.05
Absorbance (from 245 nm) ≤ 0.01
Transmittance (195 nm) ≥ 10 %
Transmittance (210 nm) ≥ 60 %
Transmittance (217 nm) ≥ 80 %
Transmittance (225 nm) ≥ 90 %
Transmittance (from 245 nm) ≥ 98 %
Fluorescence (as quinine) (254 nm) ≤ 1.0 ppb
Fluorescence (as quinine) (365 nm) ≤ 1.0 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR