1-Pentanesulphonic acid sodium salt 99, HiPerSolv CHROMANORM® HPLC

Supplier: VWR Chemicals

Synonyms: Sodium 1-pentanesulphonate, Sodium pentane-1-sulphonate, Pentane-1-sulphonic acid sodium salt, Sodium pentanesulphonate, 98%, Sodium pentanesulphonate

VWRC152812VEA 95.5 SGD
VWRC152812V VWRC152813W
1-Pentanesulphonic acid sodium salt 99, HiPerSolv CHROMANORM® HPLC
1-Pentanesulphonic acid sodium salt
Formula: C₅H₁₁O₃SNa
MW: 174.2 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007541
CAS Number: 22767-49-3
EINECS: 245-208-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddb1c23c8a6fe3c156a3aacb62721e1d1a34efb0&vid=1936dcdb47ba374ce21fd758660f7ca6ba95d254&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated on dried substance) ≥ 99,0 %
Appearance White powder
IR Spectrum Passes test
Water ≤ 2,0 %
Absorbance (200 nm) (0.25 mol/l) ≤ 0,1
Absorbance (210 nm) (0.25 mol/l) ≤ 0,05
Absorbance (220 nm) (0.25 mol/l) ≤ 0,04
Absorbance (230 nm) (0.25 mol/l) ≤ 0,03
Absorbance (240 nm) (0.25 mol/l) ≤ 0,01
Absorbance (250 nm) (0.25 mol/l) ≤ 0,01
Absorbance (260 nm) (0.25 mol/l) ≤ 0,01


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR