Trifluoroacetic acid 100%, HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals
Danger

Synonyms: TFA

Packaged under nitrogen.

VWRC153112EEA 370 SGD
VWRC153112E
Trifluoroacetic acid 100%, HiPerSolv CHROMANORM® for HPLC
Trifluoroacetic acid
Formula: C₂HF₃O₂
Boiling Pt: 71,8 °C (1013 hPa)
Melting Pt: –15 °C
Density: 1,490 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.80 %
Colour value Max. 10 APHA
Non-volatile matter Max. 0.005 %
Transmittance (210 nm) Min. 30 %
Transmittance (220 nm) Min. 60 %
Transmittance (230 nm) Min. 85 %
Transmittance (240 nm) Min. 95 %
Transmittance (250 nm) Min. 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR