Orthophosphoric acid 85% ≥85%, HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals
Danger

Synonyms: Phosphoric acid

VWRC153154DEA 93.8 SGD
VWRC153154D
Orthophosphoric acid 85% ≥85%, HiPerSolv CHROMANORM® for HPLC
Orthophosphoric acid
Formula: H₃PO₄
Melting Pt: 28 °C
Density: 1,689 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Assay Min. 85.0 %
Volatile acids Max. 0.0002 meq/g
Fe (Iron) Max. 10 ppm
Pb (Lead) Max. 2 ppm
Transmittance (254 nm) (0,1 mol/l) Min. 80 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR