Sodium chloride ≥99.9%, HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals

VWRC153274VEA 235 SGD
VWRC153274V
Sodium chloride ≥99.9%, HiPerSolv CHROMANORM® for HPLC
Sodium chloride
Formula: NaCl
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2,16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Assay (argentometric, after ignition) Min. 99.9 %
Identity Passes test
pH-value (5 %; water) 5.0 - 8.0
Transmission (206 nm; 1 mol/L; 1 cm) Min. 10 %
Transmission (254 nm; 1 mol/L; 1 cm) Min. 95 %
Br (Bromide) Max. 0.005 %
I (Iodide) Max. 0.001 %
PO4 (Phosphate) Max. 0.0005 %
SO4 (Sulphate) Max. 0.001 %
Fe (Iron) Max. 0.0001 %
K (Potassium) Max. 0.005 %
Pb (Lead) Max. 0.0002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR