Ammonia solution 32%, HiPerSolv CHROMANORM® for HPLC

Supplier: VWR Chemicals
Danger

VWRC153312KEA 111 SGD
VWRC153312K
Ammonia solution 32%, HiPerSolv CHROMANORM® for HPLC
Ammonia solution
Formula: NH₃
Boiling Pt: 32…37,7 °C (1013 hPa)
Melting Pt: –60…–57,5 °C
Density: 0,88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now

Specification Test Results

Assay 28 - 32 %
Non-volatile residue Max. 20 ppm
Fe (Iron) Max. 0.2 ppm
Pb (Lead) Max. 0.05 ppm
Transmittance (215 nm) (1 mol/l) Min. 10 %
Transmittance (254 nm) (1 mol/l) Min. 97 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR