Cerium(IV) sulphate 0.1 mol/l (0.1 N) aqueous solution, AVS TITRINORM Reag. Ph. Eur., Reag. USP Volumetric

Supplier: VWR Chemicals
Warning

Synonyms: Cerium disulphate, Ceric sulphate, Cerium(IV) sulphate

VWRC190474MEA 208 SGD
VWRC190474M VWRC190475N
Cerium(IV) sulphate 0.1 mol/l (0.1 N) aqueous solution, AVS TITRINORM Reag. Ph. Eur., Reag. USP Volumetric
Cerium (IV) sulphate
Formula: Ce(SO₄)₂
MW: 332.24 g/mol
Melting Pt: 195
MDL Number: MFCD00148852
CAS Number: 13590-82-4
EINECS: 237-029-5
UN: 3264
ADR: 8 III
Merck Index: 12,02036

Order Now

Specification Test Results

Titer (20°C; real value 0.3 % accuracy) 0,0995-0,1005 mol/l
NIST traceable Confirmed
Conforms to Reag. Ph.Eur. Passes test
Conforms to Reag. USP Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR