Vanadomolybdate reagent

Supplier: VWR Chemicals
Danger

VWRC191844WEA 206 SGD
VWRC191844W
Vanadomolybdate reagent
Vanadomolybdate reagent
Boiling Pt: ~100 °C (1013 hPa)
Density: 1,2 g/cm³ (20 °C)
UN: 2031
ADR: 8,II

Order Now

Specification Test Results

Colour reaction Satisfactory

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR