Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC

Supplier: VWR Chemicals
Danger


Filtered through a 0.2 µm filter, packaged under nitrogen.

VWRC20060.320EA 111 SGD
VWRC20060.320 VWRC20060.420 VWRC20060.290 VWRC20060.360
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Acetonitrile
Formula: H₃CCN
Boiling Pt: 81,6 °C (1013 hPa)
Melting Pt: –45,7 °C
Density: 0,782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.9 %
Acidity Max. 0.0002 meq/g
Alkalinity Max. 0.0002 meq/g
Evaporation residue Max. 5 ppm
Water Max. 0.02 %
Absorbance (195 nm) Max. 0.1
Absorbance (200 nm) Max. 0.05
Absorbance (210 nm) Max. 0.02
Absorbance (220 nm) Max. 0.02
Absorbance (240 nm) Max. 0.005
Transmittance (195 nm) Min. 80 %
Transmittance (200 nm) Min. 90 %
Transmittance (210 nm) Min. 94 %
Transmittance (220 nm) Min. 96 %
Transmittance (240 nm) Min. 99 %
Gradient grade (210 nm) Max. 10 mAU

SPECIFICATIONS

Environmentally Preferable Safer Human and Environmental Health Safer Human and Environmental Health Safer Human and Environmental Health Safer Human and Environmental Health
Environmental certification EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion Chem21 Solvent Selection Guide recommended by default and after discussion Chem21 Solvent Selection Guide recommended by default Chem21 Solvent Selection Guide recommended by default and after discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR