Acetic acid 32%, TECHNICAL

Supplier: VWR Chemicals
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

VWRC20132.364EA 195 SGD
VWRC20132.364
Acetic acid 32%, TECHNICAL
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Density: 1.04
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2790
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 32 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR