Hydrochloric acid 35%, TECHNICAL

Supplier: VWR Chemicals
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

VWRC20246.298EA 7.8 SGD
VWRC20246.298 VWRC20246.460
Hydrochloric acid 35%, TECHNICAL
Hydrochloric acid
Formula: HCl
MW: 36.46
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II
REACH: 01-2119484862-27

Order Now

Specification Test Results

Assay Min. 35 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR