Hexafluorosilicic acid 31% aqueous solution, TECHNICAL

Supplier: VWR Chemicals
Danger

Synonyms: Fluosilicic acid

VWRC20313.291EA 61.8 SGD
VWRC20313.291
Hexafluorosilicic acid 31% aqueous solution, TECHNICAL
Hexafluorosilicic acid
Formula: H₂F₆Si
MW: 144.09 g/mol
Density: 1.29 g/cm³ (20 °C)
MDL Number: MFCD00036289
CAS Number: 16961-83-4
EINECS: 241-034-8
UN: 1778
ADR: 8 II

Order Now

Specification Test Results

Assay Min. 31 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR