Formic acid 98% ≥98%, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Aminic Acid, Methanoic acid

VWRC20320.295EA 63.9 SGD
VWRC20320.295 VWRC20320.320 VWRC20320.364
Formic acid 98% ≥98%, GPR RECTAPUR®
Formic acid
Formula: HCO₂H
Boiling Pt: 100…101 °C (1013 hPa)
Melting Pt: 8,2…8,4 °C
Density: 1,22 g/cm³ (25 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8 (3),II
REACH: 01-2119491174-37
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98 %
Refractive index (20°C) 1.370 - 1.372
Acetic acid Max. 0.2 %
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 100 ppm
Evaporation residue Max. 100 ppm
Cl (Chloride) Max. 20 ppm
SO4 (Sulphate) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR