Lactic acid 90% ≥90% in aqueous solution, TECHNICAL

Supplier: VWR Chemicals
Danger

Synonyms: DL-Lactic acid, 2-Hydroxypropionic acid, 2-Hydroxypropanoic acid

VWRC20366.293EA 111 SGD
VWRC20366.293 VWRC20366.464
Lactic acid 90% ≥90% in aqueous solution, TECHNICAL
Lactic acid
Formula: H₃CCH(OH)COOH
Boiling Pt: 122 °C (20 hPa)
Melting Pt: 53 °C
Density: 1,11…1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
EINECS: 200-018-0

Order Now

Specification Test Results

Assay Min. 90 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR