L(+)-Lactic acid ≥90% Ph. Eur.

Supplier: VWR Chemicals
Danger

Synonyms: (S)-Lactic acid, L-Lactic acid, (S)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropionic acid, (S)-(+)-Lactic acid

VWRC20370.297EA 71 SGD
VWRC20370.297 VWRC20370.366
L(+)-Lactic acid ≥90% Ph. Eur.
L(+)-Lactic acid

Caution: Not for parenteral use.

Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 235
Melting Pt: 53 °C
Density: 1.11…1.21 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064266
CAS Number: 79-33-4
EINECS: 200-018-0
Merck Index: 13,05352

Order Now

Specification Test Results

Assay 88.0 - 92.0 %
Appearance Conforms (see CoA/CoS)
Identification A Passes test
Identification C Passes test
Appearance test Passes test
Ether-insoluble substances Passes test
Sugars and other reducing substances Passes test
Citric, oxalic and phosphoric acids Passes test
Relative density 1.200 - 1.210
SO4 (Sulphate) Max. 200 ppm
Ca (Calcium) Max. 200 ppm
Heavy metals (as Pb) Max. 10 ppm
Sulphated ash Max. 0.1 %
Residual solvents Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR