Mercaptoacetic acid (thioglycolic acid) ≥98%, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Mercaptoacetic acid

VWRC20495.265EA 232 SGD
VWRC20495.265
Mercaptoacetic acid (thioglycolic acid) ≥98%, GPR RECTAPUR®
Mercaptoacetic acid (Thioglycolic acid)

Caution: WARNING : The indicated concentration refers to the product leaving the factory. A continuous loss of strength, specific to the product, is predictable.

Formula: HSCH₂COOH
MW: 92.12 g/mol
Boiling Pt: 96
Melting Pt: –16,5 °C
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 130 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00004876
CAS Number: 68-11-1
EINECS: 200-677-4
UN: 1940
ADR: 8 II
Merck Index: 13,09410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45179825145ec44f2e058197a0ee54845ae7c750&vid=51e43f5b69b880b196570b3065f9b4d8db7336d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (at filling) Min. 98 %
Ignition residue (SO4) Max. 0.1 %
Fe (Iron) Max. 5 ppm
The statement of the assay applies to the test date. Agradual substance-specific content reduction, especially at higher temperatures, must be excepted.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR