Orthophosphoric acid 25%, TECHNICAL

Supplier: VWR Chemicals
Danger

Synonyms: Phosphoric acid

VWRC20629.463EA 742 SGD
VWRC20629.463
Orthophosphoric acid 25%, TECHNICAL
Orthophosphoric acid
Formula: H₃PO₄
MW: 98 g/mol
Boiling Pt: 158
Melting Pt: 21
Density: 1.1462 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8 III
Merck Index: 14,07344

Order Now

Specification Test Results

Assay 24 - 26 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR