Salicylic acid ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: 2-Hydroxybenzoic acid, o-Hydroxybenzoic acid

VWRC20655.292EA 119 SGD
VWRC20655.292
Salicylic acid ≥99%, GPR RECTAPUR®
Salicylic acid
Formula: (HO)C₆H₄CO₂H
Boiling Pt: 336 °C (1013 hPa)
Melting Pt: 157…159 °C
Density: 1,443 g/cm³ (20 °C)
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00002439
CAS Number: 69-72-7
EINECS: 200-712-3
Merck Index: 13,08411
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af1f695fa92656a135b5e1742f1cca28f4d255fb&vid=8f3aff6fb9a70e5dd194b8bc46019af74e48e90a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99 %
Melting point 158 - 161 °C
Heavy metals (as Pb) Max. 50 ppm
Ignition residue (SO4) Max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR