Stearic acid, powder, TECHNICAL

Supplier: VWR Chemicals
Warning

Synonyms: Octadecanoic acid, Octadecylic acid

VWRC20666.290EA 46.3 SGD
VWRC20666.290 VWRC20666.460
Stearic acid, powder, TECHNICAL
Stearic acid
Formula: C₁₈H₃₆O₂
MW: 284.48 g/mol
Boiling Pt: 361
Melting Pt: 67
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 196 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00002752
CAS Number: 57-11-4
EINECS: 200-313-4
REACH: 01-2119543894-28
Merck Index: 13,08882
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b000f914d1ecf0e3b0d3bc79cd318f3c7842831d&vid=b53ff33f46eb3558f286c08be23286aab54dce6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Stearic and palmitic acid Min. 90 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR