Sulphanilic acid ≥98%, GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

Synonyms: Aniline-4-sulphonic acid, 4-Aminobenzenesulphonic acid

VWRC20673.294EA 268 SGD
VWRC20673.294
Sulphanilic acid ≥98%, GPR RECTAPUR®
Sulphanilic acid
Formula: 4-(H₂N)C₆H₄SO₃H
MW: 173.19 g/mol
Boiling Pt: 363
Melting Pt: 279
Density: 1.485 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007886
CAS Number: 121-57-3
EINECS: 204-482-5
Merck Index: 13,09012
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e032bed891f24ee48419dc3c7237b0293fd3e629&vid=28238ab814bb2b8d6eb5012387d8bf41528469e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 98 %
Ignition residue (SO4) Max. 0.1 %
Cl (Chloride) Max. 100 ppm
SO4 (Sulphate) Max. 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR