Sulphuric acid ≥95%, TECHNICAL

Supplier: VWR Chemicals
Danger

VWRC20685.295EA 37.7 SGD
VWRC20685.295 VWRC20685.513 VWRC20685.330 VWRC20685.360
Sulphuric acid ≥95%, TECHNICAL
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 10.38 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II
REACH: 01-2119458838-20

Order Now

Specification Test Results

Assay Min. 95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR