Sulphuric acid ≥90% for milk analysis

Supplier: VWR Chemicals
Danger

VWRC20695.290EA 86.4 SGD
VWRC20695.290 VWRC20695.324
Sulphuric acid ≥90% for milk analysis
Sulphuric acid
Formula: H₂SO₄
Density: 1,82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II

Order Now

Specification Test Results

Assay Min. 90 %
Density (20/4) 1.815 - 1.825


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR