Isoamyl alcohol, Wasser_0,05 98, GPR RECTAPUR® zS

Supplier: VWR Chemicals
Warning

Synonyms: Isopentyl alcohol, iso-Pentanol, 3-Methyl-1-butanol, 3-Methylbutanol

VWRC20796.298EA 147 SGD
VWRC20796.298
Isoamyl alcohol, Wasser_0,05 98, GPR RECTAPUR® zS
Isoamyl alcohol
Formula: (CH₃)₂CHCH₂CH₂OH
MW: 88.15 g/mol
Boiling Pt: 130
Melting Pt: –117 °C
Density: 0.812 g/cm³ (20 °C)
Flash Pt: 42
Storage Temperature: Ambient
MDL Number: MFCD00002934
CAS Number: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3 III
Merck Index: 13,05214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=170c60e26c7b4edab6e0686f91d04e676da16e6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated as mixture of isomers) Min. 98 %
Density (20/4) 0.808 - 0.812
Water Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR